VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Toimittaja: VWR Chemicals
E504-500MLEA 61.1 EUR
E504-500ML
VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Biokemialliset puskuriliuokset Puskurit soluviljelyyn
137 mM sodium chloride, 2,7 mM potassium chloride, 12 mM phosphate buffer
  • pH 7,4