1,2-Dimetoksietaani ≥99% inhibitor-free, Reagent Grade

Toimittaja: Honeywell Chemicals
Vaara

E27408-1L E27408-2.5L E27408-20L
E27408-1LEA 143 EUR
E27408-1L E27408-2.5L E27408-20L
1,2-Dimetoksietaani ≥99% inhibitor-free, Reagent Grade
1,2-Dimetoksietaani

  • ≥95% for general laboratory use.

Kaava: C₄H₁₀O₂
Kiehumispiste: 85 °C (1,013 hPa)
Sulamispiste: -58 °C
Tiheys: 0,87 g/cm³ (20 °C)
Leimahduspiste: -2 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008502
CAS-numero: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II
Merck Index: 13,03251

Order Now

Testitulosten erittely

Identity (IR) complying
Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 0.5 %
Peroxides (as H₂O₂) Max. 15 ppm
Vapor Pressure 48 mmHg ( 20 °C)
Explosion Limit 10.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR