Suojahaalari, kemikaalinkestävä, Tyvek® 600 Plus
Toimittaja: DuPont
TYCHA5TWHSM010 TYCHA5TWHMD010 TYCHA5TWHLG010 TYCHA5TWHXL010 TYCHA5TWH2X010 TYCHA5TWH3X010 TYCHA5TWHLG008 TYCHA5TWHMD008 TYCHA5TWHSM008 TYCHA5TWHXL008 TYCHA5TWH2X008 TYCHA5TWH3X008 TYCHA5TGRMD010 TYCHA5TGRLG010 TYCHA5TGR2X010 TYCHA5TGRXL010 TYCHA5TWH4X010 TYCHA5TWH7X010 TYCHA5TWH5X010 TYCHA5TWH6X010 TYCHA5TGRSM010 TYCHA5TGR5X010 TYCHA5TGR6X010 TYCHA5TGR7X010 TYCHA5TGR3X010 TYCHA5TGR4X010 TYCHA5TWHXS010 TYCHA5TWH4X008 TYCHA5TWH5X008 TYCHA5TWH6X008 TYCHA5TWH7X008 TYCHA5TWHXS008 TYCHA5TGRXS010
113-3874EA 367 EUR
113-3874 113-3875 113-3876 113-3877 113-3878 113-3879 113-3894 113-3893 113-3892 113-3895 113-3896 113-3897 113-9701 113-9702 113-9704 113-9703 DUPOTYCHA5TWH4X010 DUPOTYCHA5TWH7X010 DUPOTYCHA5TWH5X010 DUPOTYCHA5TWH6X010 DUPOTYCHA5TGRSM010 DUPOTYCHA5TGR5X010 DUPOTYCHA5TGR6X010 DUPOTYCHA5TGR7X010 DUPOTYCHA5TGR3X010 DUPOTYCHA5TGR4X010 DUPOTYCHA5TWHXS010 DUPOTYCHA5TWH4X008 DUPOTYCHA5TWH5X008 DUPOTYCHA5TWH6X008 DUPOTYCHA5TWH7X008 DUPOTYCHA5TWHXS008 DUPOTYCHA5TGRXS010
Suojahaalari, kemikaalinkestävä, Tyvek® 600 Plus
Kertakäyttöiset vaatteet
These Tyvek® overalls are made from flash spun, high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size).
  • Ultra-low linting fabric is robust yet lightweight (<250 g per garment)
  • With hood; elasticated face, wrists and ankles (tunnelled), glued-in waist elastic
  • Tyvek® zip and zip flap for enhanced protection
  • Self-adhesive chin flap for