Chemical resistant overalls, Tyvek® 800 J, model TJ 198 Ta
Toimittaja: DuPont
TJ198TWHXL0025 TJ198TWH3X0025 TJ198TWHSM0025 TJ198TWHMD0025 TJ198TWHLG0025 TJ198TWH2X0025 TJ198TWH4X0025 TJ198TWH5X0025 TJ198TWH6X0025 TJ198TWH7X0025
DUPOTJ198TWHXL0025EA 469 EUR
DUPOTJ198TWHXL0025 DUPOTJ198TWH3X0025 DUPOTJ198TWHSM0025 DUPOTJ198TWHMD0025 DUPOTJ198TWHLG0025 DUPOTJ198TWH2X0025 DUPOTJ198TWH4X0025 DUPOTJ198TWH5X0025 DUPOTJ198TWH6X0025 DUPOTJ198TWH7X0025
Chemical resistant overalls, Tyvek® 800 J, model TJ 198 Ta
Kertakäyttöiset vaatteet
This robust yet lightweight garment benefits from the use of a specific 'impervious technology' to achieve an effective barrier against many low-concentrated water-based inorganic chemicals (even under pressure) and small-sized hazardous particulates, as well as oil repellences.
  • Anti-static treatment on inside
  • Robust yet lightweight (<300 g)
  • Hood, Tyvek® zip and self-adhesive zip flap for enhanced protection
  • Self-adhesive chin flap for tight seal of suit to mask
  • Elasticated face, wrists and ankles as well as glued-in waist elastic for good fit
  • Thumb
More Product Information
Order Now
TEKNISET TIEDOT
Tyyppi 800 J, TJ 198 Ta
Environmentally Preferable
Antisaattisuus +
Sukupuoli
Asu sovellukset Chemical protection,Laboratory,Industrial
Paino 59 g/m²