Safety cabinets for pressurised gas cylinders, type G30, ECO line plus
Toimittaja: DUPERTHAL
73-200960-011 73-201260-011 73-200660-011
DUEH73-200960-011EA 2990 EUR
DUEH73-200960-011 DUEH73-201260-011 DUEH73-200660-011
Safety cabinets for pressurised gas cylinders, type G30, ECO line plus
Kalusteet Säilytyskaapit
Safety cabinets for pressurised gas cylinders-inside storage is made up of sheet steel, powder-coated.
  • Classical door technology
  • Access control with cylinder lock
  • Enhanced ventilation in every cabinet level
  • Adjustable feet for level equalisation
Sertifikaatit: Conforms with DIN EN 14470-2, EK5/AK4 09-10, DIN EN 14727, CE certified, TRGS 510 chapter 10, type G30.