Trolleys, gas cylinders
Toimittaja: DUPERTHAL
7.00.600 7.00.620 7.00.630 7.01.000 7.01.010 7.01.020 7.01.030 7.01.035
DUEH7.00.600EA 236 EUR
DUEH7.00.600 DUEH7.00.620 DUEH7.00.630 DUEH7.01.000 DUEH7.01.010 DUEH7.01.020 DUEH7.01.030 DUEH7.01.035
Trolleys, gas cylinders
Kuljetuskärryt, siivousvaunut
These trolleys constructed of blue painted steel tube are ideal for handling gas cylinders.
  • With solid rubber tyres
  • Includes secure chain
  • Finger guards for safe grip