Pipette controller, Pipet-Aid® XP2
Toimittaja: DRUMMOND
4-000-501-I 4-000-501-UK
DRUM4-000-501-IEA 1620 EUR
DRUM4-000-501-I DRUM4-000-501-UK
Pipette controller, Pipet-Aid® XP2
Pipetit ja pipetintäyttäjät Pipetintäyttäjät
This controller is an advanced version of the Pipet-Aid® XP controller and accepts 1 to 100 ml pipettes.
  • New ergonomic design
  • New power source for uninterrupted extended operation; can be charged while in use
  • Ultra quiet precision pump provides great control for aspiration or dispensing
Toimitustiedot: The unit is supplied complete with a power supply/charger, four extra filters, and a removable stand.