Microinjectors, Nanoject III™
Toimittaja: DRUMMOND
3-000-207
DRUM3-000-207EA 4690 EUR
DRUM3-000-207
Microinjectors, Nanoject III™
Mikroinjektorit Mikroinjektori, ohjelmoitava
The Nanoject III™ microinjector is designed to perform single manual injections and multiple injection cycle recipes. The auto homing feature eliminates the need to rehome the unit and reduce jamming of the plunger.
  • User-friendly programmable touchscreen operation
  • Accurate injection volumes down to 0,6 nanoliters
  • Ensures accurate and consistent injection volumes with hydraulic technology
  • O-rings not required to secure the micropipette
  • Lower injection volume capability
Toimitustiedot: Supplied with 4 interchangeable adapter plugs.