Gas detectors, Dräger X-am® 5000
Toimittaja: Dräger
8320000 6810881 6810883 6810885 6810889 6811525 6810882 6810884 6811410 6812950 6810888 6812370 6810890
DRAE8320000EA 0 EUR
DRAE8320000 DRAE6810881 DRAE6810883 DRAE6810885 DRAE6810889 DRAE6811525 DRAE6810882 DRAE6810884 DRAE6811410 DRAE6812950 DRAE6810888 DRAE6812370 DRAE6810890
Gas detectors, Dräger X-am® 5000
Kaasun havaitsemisjärjestelmät
This gas detection system without sensor reliably measures combustible gases and vapours as well as oxygen, harmful concentrations and organic vapours.
  • Practical design and light weight make it comfortable to carry
  • Dependable warnings in the event of explosive atmospheres
  • Pump to use the detector for pre-entry measurements in confined spaces (optional)