Airlock Tray Handle
Toimittaja: DON WHITLEY SCIENTIFIC
A09422
DOWIA09422EA 0 EUR
DOWIA09422
Airlock Tray Handle
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
A useful handle for use with Whitley airlock trays. This tool gives you additional reach to pull trays from the airlock, move them around inside the workstation and push trays back into the airlock.