MiniVacuumBox 16 L blue
Toimittaja: DELTA T
B100116
DLTTB100116EA 804 EUR
DLTTB100116
MiniVacuumBox 16 L blue
Laatikot Kuljetuslaatikot
Suitable for 5 pairs of TempShell elements or combinations of elements and TempShell frames.
Sealable with double zipper, plastic interior, strap, documents bag, equipped with vacuum insulation.