TASKI® Sprint E3c Glass and Multi Purpose Cleaner
Toimittaja: Diversey
7513049
DIVY7513049EA 38.7 EUR
DIVY7513049
TASKI® Sprint E3c Glass and Multi Purpose Cleaner
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
Glass and multi purpose cleaner for the streak-free cleaning of glass, mirrors and other high gloss surfaces.
  • Highly effective, pH neutral at in-use dilution
  • Fast acting, fast drying
  • Unique, patented Odour Neutralizing Technology (O.N.T.)
  • Fresh fragrance
  • Dries quickly without streaking
Suitable for use on various water-resistant hard surfaces, e.g. windows, mirrors, glazed ceramic as well as plastic surfaces.