Lab markers, Teeny Writers
Toimittaja: DIVERSIFIED BIOTECH
TNY-WTR TNY-TWIN
DIVETNY-WTREA 17.9 EUR
DIVETNY-WTR DIVETNY-TWIN
Lab markers, Teeny Writers
Kynät Lab Markers
A retractable Sharpie® pen with an extra fine tip. Recommended for writing on all our labels for samples that will be stored at room temperature and not exposed to chemicals and/or caustic agents.