Cooling systems, CoolSafe™
Toimittaja: DIVERSIFIED BIOTECH
CSAF-1000 CSAF-2000 CSAF-3000 CSAF-4000 CHAM-1000 CHAM-2000 CHAM-3000 CHAM-4000 BRIK-1520 BRIK-2501 BRIK-1501 CSF-BOX CSAF-3050 CSAF-5000 CHAM-3050 CHAM-5000 CHAM-1020 CHAM-3840 CHAM-8000 CHAM-7050
DIVECSAF-1000EA 984 EUR
DIVECSAF-1000 DIVECSAF-2000 DIVECSAF-3000 DIVECSAF-4000 DIVECHAM-1000 DIVECHAM-2000 DIVECHAM-3000 DIVECHAM-4000 DIVEBRIK-1520 DIVEBRIK-2501 DIVEBRIK-1501 DIVECSF-BOX DIVECSAF-3050 DIVECSAF-5000 DIVECHAM-3050 DIVECHAM-5000 DIVECHAM-1020 DIVECHAM-3840 DIVECHAM-8000 DIVECHAM-7050
Cooling systems, CoolSafe™
Laatikot Pöytämalliset jäähdyttimet
Polystyrene cool safe box, aluminium cooling chamber.
  • Durable
  • Temperature stable
  • Corrosion resistant
Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.