Single-use bioreactor systems, BIOne
Toimittaja: DISTEK
2022-1001 2022-1002 2022-1014 2022-1015 2022-1018 2022-1021 2022-1024 2022-1025 2022-1028 2022-1029 2022-1005 2022-1006 2022-1016 2022-1017 2022-1019 2022-1022 2022-1023 2022-1026 2022-1027 2022-1030 2022-1031 2022-1034 2022-1035 2022-1036 2022-1038 2022-1040 2022-1056 2022-1057
DISI2022-1001EA 0 EUR
DISI2022-1001 DISI2022-1002 DISI2022-1014 DISI2022-1015 DISI2022-1018 DISI2022-1021 DISI2022-1024 DISI2022-1025 DISI2022-1028 DISI2022-1029 DISI2022-1005 DISI2022-1006 DISI2022-1016 DISI2022-1017 DISI2022-1019 DISI2022-1022 DISI2022-1023 DISI2022-1026 DISI2022-1027 DISI2022-1030 DISI2022-1031 DISI2022-1034 DISI2022-1035 DISI2022-1036 DISI2022-1038 DISI2022-1040 DISI2022-1056 DISI2022-1057
Single-use bioreactor systems, BIOne
Bioreaktorit
Distek, Inc. has developed a bench top scale single-use bioreactor (SUB) for mammalian cell growth, cell therapy, insect cells and other unique applications performed in a stirred tank bioreactor. The gamma irradiated BIOne SUB is engineered with a disposable head plate welded to a triple-layered liner that can be easily inserted into a non sterile bioreactor glass vessel, converting it to a disposable SUB within a matter of seconds.
  • Customisable above and below the head plate
  • 2 and 5 L working volume
  • Optional single-use pH probe
  • Increased throughput