Microscope objective lens mounts
Toimittaja: DIDALAB
POD060550 POD060560
DIDAPOD060550EA 98.9 EUR
DIDAPOD060550 DIDAPOD060560
Microscope objective lens mounts
Mikroskoopin objektiivit
The objective holder allows precise positioning of a microscope objective according to the vertical and the horizontal.
  • Both axes 2 mm each
  • Holder is mounted on a 10 mm diameter rod