Lämpömittari, hälyttävä lämpömittari, MM120
Toimittaja: DICKSON
MM120
DICKMM120EA 151 EUR
DICKMM120
Lämpömittari, hälyttävä lämpömittari, MM120
Lämpömittarit Digital Thermometers
The MM120 is a simple, cost-effective temperature monitoring tool. The MM120 does not track your data, but it is a useful backup to a traditional data logger in your vaccine or drug monitoring application.
  • Simple, cost effective temperature monitoring too
  • Tamper-resistant audible/visual alarms
  • Thermistor probe submersed in glass bead solution for stabilised readings
  • Tracks minimum/maximum temperatures
  • Easy to read display
Ideal as a backup device to data loggers that record temperature values over time. The FDA and CDC specifically recommend usingMore Product Information