Brij® 58 10% in aqueous solution, Proteomics Grade

Toimittaja: G Bioscienes

Synonyms: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether

DG005 DG006
DG005EA 392 EUR
DG005 DG006
Brij® 58 10% in aqueous solution, Proteomics Grade
Brij® 58

G-Biosciences’ Proteomic Grade Detergent Solutions contain reduced peroxides and carbonyl compounds. In addition, the detergents have less than 50µS conductivity. These detergents are offered as sterile, 10% aqueous solutions, sealed under inert gas, and are suitable for protein applications. These non-ionic detergents are suitable for isolating membrane-protein complexes.

Kaava: C₅₆H₁₁₄O₂₁
MW: 1123.51 g/mol
MDL numero: MFCD00080892
CAS-numero: 9004-95-9
EINECS: 500-014-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR