Inoculating loops, PS, calibrated
Toimittaja: DELTALAB
302753 302743 302744 302754
DELT302753EA 312 EUR
DELT302753 DELT302743 DELT302744 DELT302754
Inoculating loops, PS, calibrated
Viljelysauvat mikrobiologiaan Viljelysilmukat
These calibrated, dual use inoculating loops offer a loop at one end to facilitate microbiological streaking, and a fine tip at the other specially designed for counting colonies.
  • High robustness and flexible
  • Colour-coded for ease of size identification
  • Eliminates the need for flaming
  • Sterilised by radiation