Bottles, wide neck, PE, rectangular, with screw cap
Toimittaja: DELTALAB
292820G
DELT292820GEA 399 EUR
DELT292820G
Bottles, wide neck, PE, rectangular, with screw cap
Pullot Muovipullot
PE, translucent, with red screw cap.
  • Good chemical resistance
  • Godd temperature resistance
  • Graduated