Visor, Visor-Toric
Toimittaja: DELTA PLUS
VISORTOIN
DELPVISORTOINEA 31.3 EUR
DELPVISORTOIN
Visor, Visor-Toric
Kasvosuojaimet
Clear polycarbonate injected visor with electric arc protection. Suitable for face shield holder VISOR HOLDER and VIROR H (BALBI2 and PICO2 support). Ideal for forest work, building and construction and cutting applications.
  • Universal fixing
  • Protection against large solid particles
  • Easy to use and comfortable
Thickness: 1,9/2,0 mm
Weight: 162 g
Sertifikaatit: EN 166, EN 170 (2C-1,2)
Tilaustiedot: The face shield holder is not included and needs to be ordered separately. Please contact your local Avantor office for further details.