Powered air purifying respirators (PAPR), Junior B series
Toimittaja: HONEYWELL SAFETY
A114201 A114200 A114400
DALLA114201EA 1350 EUR
DALLA114201 DALLA114200 DALLA114400
Powered air purifying respirators (PAPR), Junior B series
Hengityssuojaimet Hengityssuojajärjestelmät
Heavy duty air hoods range designed for use with powered air purifying respirator (PAPR) as Compact Air 200, Compact Air and Compact Air. This model offers protection to the head, the shoulders and the neck. It is equipped with full safety helmet. This system procures air to the user via the supply hose connected between PAPR (equipped with filters) and the hood. The filtered airflow enters by the front of the hood and leaves the hood via the face collar or the neck bib.
  • Soft washable neck bib with elastic security cord and unique system of spring-loaded