E.Z.N.A.® Endo-Free Plasmit Kit -pakkaukset

Toimittaja: OMEGA BIO-TEK

D6926-03 D6926-04 D6915-03 D6915-00 D6926-00
D6926-03EA 570 EUR
D6926-03 D6926-04 D6915-03 D6915-00 D6926-00
E.Z.N.A.® Endo-Free Plasmit Kit -pakkaukset
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents

Plasmid isolated with traditional purification procedures normally contain high levels of endotoxins that can significantly interfere with transfection experiments downstream. The E.Z.N.A.® Endo-Free Plasmid kits integrate an efficient endotoxin removal step into the plasmid purification procedure to produce high quality transfection grade plasmid.

Order Now

TEKNISET TIEDOT

Kuvaus E.Z.N.A.® Fastfilter® Endo-Free Plasmid Maxi kit E.Z.N.A.® Endo-Free Plasmid Midi Kit E.Z.N.A.® Fastfilter® Endo-Free Plasmid Maxi Kit
Environmentally Preferable
Hakemus
Target molecule Plasmid Plasmid Plasmid
Refine target molecule Plasmid (endotoxin-free) Plasmid (endotoxin-free) Plasmid (endotoxin-free)
Sample type Bacteria Bacteria Bacteria
Method / Format Column (silica-based) Column (silica-based) Column (silica-based)
Prep size Maxi Midi Maxi
Näytteen koko 50 - 200 ml 15 - 50 ml 50 - 200 ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR