E.Z.N.A.® Soil DNA kit
Toimittaja: OMEGA BIO-TEK
D5625-00 D5625-01 D5625-02
D5625-00EA 25.5 EUR
D5625-00 D5625-01 D5625-02
E.Z.N.A.® Soil DNA kit
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® soil DNA kit is formulated to isolate high purity cellular DNA from soil samples typically containing humic acid and inhibitors of PCR. This kit has been successfully used to isolate DNA from tough-to-lyse bacteria, fungi, and algae that inhabit a range of samples including clay, sandy, peaty, chalky, and loamy soil samples. Isolated DNA can be used for most downstream applications including PCR, southern blotting, and SNP analysis.
  • DNA isolation in 1 hour
  • Optimised buffers guarantee pure DNA every time
  • Purified DNA suitable for most downstream applications