Forensic DNA kit, E.Z.N.A.®
Toimittaja: OMEGA BIO-TEK
D3591-00 D3591-02
D3591-00EA 31.3 EUR
D3591-00 D3591-02
Forensic DNA kit, E.Z.N.A.®
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Forensic DNA Kit is designed to provide a rapid and easy method for the isolation of genomic DNA from forensic samples such as dry blood, buccal swabs, and semen samples for consistent PCR and Southern analysis. This kit can also be used for the preparation of genomic DNA from mouse tail snips, whole blood, buffy coat, serum, and plasma.
  • Process single or multiple samples simultaneously
  • No need for phenol/chloroform extractions
  • Time-consuming steps such as precipitation with isopropanol or ethanol are eliminated
DNA purified using the E.Z.N.A.® Forensic DNA Kit is ready for applications such as PCR, Southern blotting, and restriction enzyme digestion.
Sample size: 30 mg tissue or 107 cells