Hopeadietyyliditiokarbamaatti, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY
Varoitus

D3132-25G D3132-5G
D3132-25GEA 281 EUR
D3132-25G D3132-5G
Hopeadietyyliditiokarbamaatti, Sigma-Aldrich®
Hopeadietyyliditiokarbamaatti

Spin trap used in conjunction with Fe²⁺ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO₂ has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes. Also used for colorimetric determination of small quantities of arsenic. Silver diethyldithiocarbamate has been used in cell culture. Also used for colorimetric determination of small quantities of arsenic. Spin trap used in conjunction with Fe²⁺ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO₂ has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.


  • Inhibits induction of macrophage nitric oxide synthase
  • Silver diethyldithiocarbamate (AgDDC) is used to estimate the level of arsenic in human
  • Inhibits induction of macrophage nitric oxide synthase

Kaava: (C₂H₅)₂NCSSAg
Sulamispiste: 172…175 °C (kirj.)
Storage Temperature: Jääkaappi
MDL numero: MFCD00004929
CAS-numero: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c07412a6ac577902b87e980dc94c68b5023241ea&vid=1008727afdc6133fab6eb361e7594c49f93728c2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR