Supelco® TraceCERT® Organic Reference Standard, Acetochlor
Toimittaja: Merck
CRM69174-100MG
CRM69174-100MGEA 139 EUR
CRM69174-100MG
Supelco® TraceCERT® Organic Reference Standard, Acetochlor
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P201 - P273 - P280 - P302 + P352 - P304 + P340 + P312 - P308 + P313