Supelco® TraceCERT® Organic Reference Standard, Indoxacarb
Toimittaja: Merck
CRM67344-25MG
CRM67344-25MGEA 137 EUR
CRM67344-25MG
Supelco® TraceCERT® Organic Reference Standard, Indoxacarb
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P273 - P280 - P301 + P310 - P302 + P352 - P304 + P340 + P312 - P314