Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb-sulfoxide
Toimittaja: Merck
CRM05148-10MG
CRM05148-10MGEA 168 EUR
CRM05148-10MG
Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb-sulfoxide
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P264 - P270 - P301 + P310 - P405 - P501