Comfort belt style fall arrest harness, Protecta®, 3M™
Toimittaja: CAPITAL SAFETY
1161627 1161628 1161629 1161633 1161634 1161635 1161645 1161646 1161647
CPSD1161627EA 0 EUR
CPSD1161627 CPSD1161628 CPSD1161629 MMMA1161633 MMMA1161634 MMMA1161635 CPSD1161645 CPSD1161646 CPSD1161647
Comfort belt style fall arrest harness, Protecta®, 3M™
Putoamissuojat Putoamissuojaimet, turvavaljaat
The Protecta® Comfort Belt Style Fall Arrest Harness is designed to provide reliable protection, comfort, and convenience. It now features upgraded features and a sleek, modern look.
  • Stain and water resistant webbing keeps the harness cleaner and dryer
  • Steel, zinc-plated black electro-deposition paint D-rings
  • Conveniently placed front D-ring helps users stay balanced and centred
  • Auto-resetting lanyard keeper provides easy parking and release of snap hooks or carabiners
  • Impact indicators make it simple to inspect the harness for damage and help ensure it’s