Fall protection harness accessories, Protecta®, 3M™
Toimittaja: CAPITAL SAFETY
1150491 1150494
CPSD1150491EA 0 EUR
CPSD1150491 CPSD1150494
Fall protection harness accessories, Protecta®, 3M™
Putoamissuojat Putoamissuojaimet, turvavaljaat
Protecta® fall arrest harness accessory options provide the opportunity to add extra tool carrying convenience and comfort to a harness.
  • Tools can be secured while working at heights, to keep people and property safe anywhere below
  • Suitable for use with Protecta® Comfort belt harness models only
The Protecta® Belt loop D-ring keeps tools secured while working at height.
The Protecta® Comfort shoulder/back padding and comfort leg padding increase wearer comfort while in harness.