Rod baffles
Toimittaja: COWIE
PC708.012.250 PC708.008.150 PC708.008.200 PC708.008.250 PC708.012.200 PC708.016.150 PC708.016.200 PC708.016.250 PC708.012.150
COWIPC708.012.250EA 186 EUR
COWIPC708.012.250 COWIPC708.008.150 COWIPC708.008.200 COWIPC708.008.250 COWIPC708.012.200 COWIPC708.016.150 COWIPC708.016.200 COWIPC708.016.250 COWIPC708.012.150
Rod baffles
Sekoittajat Sekoitustyökalut
Rod baffles featuring a stainless steel core fully encapsulated in PTFE.
  • Used to increase agitation and to promote thorough mixing
  • Chemical resistant and non-contaminating
  • For assembly with corresponding compression fitting