Sleeves for conical ground joints
Toimittaja: COWIE
019.010 019.012 019.014 019.019 019.024 019.029 019.034 019.040 019.045 019.050 019.055 019.019.1 019.014.1
COWI019.010EA 8.6 EUR
COWI019.010 COWI019.012 COWI019.014 COWI019.019 COWI019.024 COWI019.029 COWI019.034 COWI019.040 COWI019.045 COWI019.050 COWI019.055 COWI019.019.1 COWI019.014.1
Sleeves for conical ground joints
NS-liitosholkit Liitosholkit
Thin wall joint sleeves, made of PFTE, are inserted between standard taper ground glass joints to eliminate the use of grease and prevent sticking.
  • Sleeves have a wall thickness of about 0.05mm are inert
  • Usable to 280˚C
  • Withstand low to medium vacuum