Standard valves
Toimittaja: COWIE
016.1203.2 016.1204.2 016.1302.6.2 016.1303.2 016.1304.2 016.804.2 016.803.2 016.802.5.2 016.802.2 016.704.2 016.703.2 016.702.5.2 016.702.2 016.703.7.2 016.704.9.2 016.803.7.2 016.804.9.2 016.705.2 016.805.2 016.1202.6.2
COWI016.1203.2EA 64.1 EUR
COWI016.1203.2 COWI016.1204.2 COWI016.1302.6.2 COWI016.1303.2 COWI016.1304.2 COWI016.804.2 COWI016.803.2 COWI016.802.5.2 COWI016.802.2 COWI016.704.2 COWI016.703.2 COWI016.702.5.2 COWI016.702.2 COWI016.703.7.2 COWI016.704.9.2 COWI016.803.7.2 COWI016.804.9.2 COWI016.705.2 COWI016.805.2 COWI016.1202.6.2
Standard valves
Hanat
Standard valves made of PTFE.
  • Maximum Pressure 1 bar at 100 °C and 5 mm Hg
  • Valves may be sterlised at 135 °C
  • Very good chemical resistance