Stirrer guides, universal, PTFE
Toimittaja: COWIE
005.202.7 005.206.7 005.201.7 005.204.7 005.203.7 005.205.7 005.207.7 005.208.7 005.209.7 005.211.7 005.212.7 PC709.0825 PC709.0819 PC709.1019 PC709.1025 PC709.1219 PC709.1225 PC709.1625 005.112.7 005.111.7 005.108.7 005.106.7 005.105.7 005.104.7 005.102.7 005.101.1.7
COWI005.202.7EA 605 EUR
COWI005.202.7 COWI005.206.7 COWI005.201.7 COWI005.204.7 COWI005.203.7 COWI005.205.7 COWI005.207.7 COWI005.208.7 COWI005.209.7 COWI005.211.7 COWI005.212.7 COWIPC709.0825 COWIPC709.0819 COWIPC709.1019 COWIPC709.1025 COWIPC709.1219 COWIPC709.1225 COWIPC709.1625 COWI005.112.7 COWI005.111.7 COWI005.108.7 COWI005.106.7 COWI005.105.7 COWI005.104.7 COWI005.102.7 COWI005.101.1.7
Stirrer guides, universal, PTFE
Sekoittajat Sekoitustyökalut
Stirrer guide features composite PTFE/PEEK seal and a glass ball-bearing for rigidity and smoothness of operation.
  • Resistant to chemicals
  • Anti-whip and reduced vibration
They have an NPT screw thread fitting for direct assembly into PTFE reactor lids or NPT extension adapters and standard taper cone fittings are for use in standard glass joints.