SARS-CoV-2 Antigen rapid test cassette (nasal swab)
Toimittaja: SURESCREEN
COVID19AGVCG COVID19AGVCGEU
COVID19AGVCGEA 272 EUR
COVID19AGVCG SURECOVID19AGVCGEU
SARS-CoV-2 Antigen rapid test cassette (nasal swab)
Bio-assayt Immunologiset analyysit
A rapid test for the qualitative detection of SARS-CoV-2 antigen in a nasal swab.
INTENDED USE:
The SARS-CoV-2 antigen rapid test cassette is a rapid chromatographic immunoassay for the qualitative detection of SARS-CoV-2 antigen in a Nasal swab. The identification is based on monoclonal antibodies specific to the Nucleocapsid (N) protein of SARS-CoV-2. It is intended to aid in the rapid differential diagnosis of COVID-19 infection.
Varoitus: For professional in vitro diagnostic use only.