Hazardous products shippers
Toimittaja: E3 Cortex
FRIGIPACK4L
CORTFRIGIPACK4LEA 156 EUR
CORTFRIGIPACK4L
Hazardous products shippers
Lähetys- ja kuljetuslaatikot
Isothermal packaging allowing for shipment at low temperature of products sensitive to temperature variations.
  • Available in standard version
  • Supplied with preassembled isothermal barrier in 60 mm PSE