10 chamber with vent caps, Corning® CellSTACK®
Toimittaja: Corning
7073
CORN7073EA 1540 EUR
CORN7073
10 chamber with vent caps, Corning® CellSTACK®
Bioreaktorit
Has 6360 cm² cell growth area, two 26 mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system, Standard 33 mm threaded caps have 0,2 µm pore nonwettable membranes sealed directly to the caps to allow gas exchange while minimising the risk of contamination, Optional 33 mm threaded caps are available with integrally sealed chemically resistant.
Heat sealable flexible tubing to allow direct sterile transfer of media and cells, USP Class VI polystyrene providesMore Product Information