Digital dry bath heaters, LSE™ Corning®
Toimittaja: Corning
6886-DB 6876-SB 6896-FB
CORN6886-DBEA 701 EUR
CORN6886-DB CORN6876-SB CORN6896-FB
Digital dry bath heaters, LSE™ Corning®
Kuivahauteet Digitaaliset kuivahauteet
The Corning Digital Dry Bath Heaters are available in single, dual, and four block configurations and provide a broad temperature range up to 150 °C, which makes them useful for a variety of applications in molecular biology, histology, clinical, environmental, and industrial laboratories.
  • Single, dual, and four block capacity to maximize sample processing
  • Dual display allows for easy monitoring of temperature and time
  • USB connectivity enables traceability of data
  • Molded block chamber provides excellent temperature uniformity
Corning® LSE™ Digital DryMore Product Information