Volumetric flasks, class B, with tapered stopper, Corning®
Toimittaja: Corning
5641P-100 5641P-1L 5641P-50 5641P-500 5641P-10 5641P-250
CORN5641P-100EA 177 EUR
CORN5641P-100 CORN5641P-1L CORN5641P-50 CORN5641P-500 CORN5641P-10 CORN5641P-250
Volumetric flasks, class B, with tapered stopper, Corning®
Kolvit ja erlenmeyerit Mittapullot
Made of PP, translucent, with tapered PP stopper.
  • With an individually calibrated graduation line
  • Recommended routine cleaning up to 60 °C will preserve graduations
  • Exposure to temperatures above 60 °C may affect accuracy
Reusable plastic flasks are manufactured to Class B tolerances as defined by ISO 1042.
The accuracy of volumetric ware depends on the care used in calibrating it, using the correct type of ware for the application, handling the ware properly and ensuring the ware is clean. Calibration of volumetric ware is usually done at 20 °CMore Product Information