96-Well high content screening microplates with film bottom, Corning®
Toimittaja: Corning
4680 4517
734-2763EA 400 EUR
734-2763 CORN4517
96-Well high content screening microplates with film bottom, Corning®
Kuoppalevyt Levyt suurtehoseulontaan
With an ultraclear film bottom and excellent flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications.
  • TC-treated, flat well bottom black microplate
  • Made of cyclic olefin copolymer
  • Ultraclear 190 µm film bottom thickness for imaging microscopy applications
  • Low background fluorescence and minimal crosstalk allows highest possible optical quality for cell based assays
  • Half area 96-well microplate reduces reagent consumption
  • Sterile
  • Inter and inter-well film bottom flatness optimised for high content