Disposable aseptic transfer cap for disposable spinner flask, Corning®
Toimittaja: Corning
3562 3564 3565 3567 3558 3545 3528
734-4098EA 457 EUR
734-4098 734-4100 734-4101 734-4102 CORN3558 CORN3545 CORN3528
Disposable aseptic transfer cap for disposable spinner flask, Corning®
Tulpat ja korkit Pullojen sulkimet
These disposable Corning® spinner flask aseptic transfer caps will both reduce contamination and enhance the productivity of your cell culture operations when adding or removing medium and cells.
  • Cap is made of high density polyethylene; autoclaving is not recommended
  • The venting port ends in a 25 mm diameter vent filter with a 0,2 µm pore PVDF membrane and a barbed fitting
  • Chemically resistant, heat sealable flexible tubing, ending in a male medical plastic coupler (MPC) connector closed with a female sealing cap
Aseptic transfer caps are sterile (10⁻More Product Information