Avantor® ACE® UltraCore C18, Analytical HPLC/UHPLC Columns
Toimittaja: Avantor
CORE-35F-521EA 652 EUR
CORE-35F-521 CORE-35F-530 CORE-35F-546 CORE-35F-1021 CORE-35F-1030 CORE-35F-1046 CORE-35F-1521 CORE-35F-1530 CORE-35F-1546 CORE-35F-21GD CORE-35F-30GD CORE-35F-46GD
Avantor® ACE® UltraCore C18, Analytical HPLC/UHPLC Columns
Kromatografiakolonnit
Avantor® ACE® UltraCore C18 are high performing solid-core (core-shell) particle columns designed for a wide range of LC applications. This densely bonded octadecyl phase can be used to separate acidic, basic and neutral molecules, based on hydrophobic retention mechanisms.
  • High purity, base deactivated silica with excellent reproducibility
  • High efficiency separations comparable with sub-2 µm packings but lower back pressure
  • Ideal for a wide range of analytes differing in hydrophobicity
These stainless steel columns are available in a variety ofMore Product Information