Storage Boxes, Rectangular
Toimittaja: Gilac PROFESSIONNEL
G119921 G118921 G118977 G119621 G119677 G119821 G119877 G119977 G193377
CORAG119921EA 67 EUR
CORAG119921 CORAG118921 CORAG118977 CORAG119621 CORAG119677 CORAG119821 CORAG119877 CORAG119977 CORAG193377
Storage Boxes, Rectangular
Laatikot Säilytyslaatikot
These rectangular storage boxes made from HDPE are ideal for sub-zero and ambient temperature storage and transporting raw materials.
  • No gaps or overlaps, rounded corners for easy cleaning
  • Machine washable
  • Optional lid for improved hygiene
  • Stackable with and without lid
  • Vertical lateral reinforcements for increased load support
  • Compatible with Gilac 4- and 6-wheel dollies
Tilaustiedot: Lids need to be ordered separately.