PROSAT® QuickTask™ Mopping System
Toimittaja: CONTEC
PSTA0021-BPR PSTA0022-BPR
CONTPSTA0021-BPREA 479 EUR
CONTPSTA0021-BPR CONTPSTA0022-BPR
PROSAT® QuickTask™ Mopping System
Siivousvälineet
Mops presaturated with USP Grade IPA provide consistency between operators and eliminate the need to take extra cleaning solution and separate mops and buckets into a room. Presaturated mops allow for targeted application of IPA to surfaces. They eliminate the risk of flammable liquid spills and reduce IPA usage and VOCs. Presaturated mops also provide an ergonomic option to remove residues.
  • IPA presaturated QuickTask™ mop
  • 100% 2-ply polyester covered mop heads release low levels of particles and fibres
  • Available irradiated, making it ideal for use in