Cleanroom wipes, Anticon Guardrail® G2
Toimittaja: CONTEC
G2HW0101
CONTG2HW0101EA 939 EUR
CONTG2HW0101
Cleanroom wipes, Anticon Guardrail® G2
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
100% continuous filament polyester wipe with sealed border and Particle Attraction Technology (PAT).
  • Sealed border edge allows wipe to have low fibers and particles
  • Highly absorbent and abrasion resistant
  • Particle Attraction Technology (PAT) attracts 35 times more particles than other wipes
  • Excellent solvent compatibility with isopropyl alcohol, acetone, and other solvents
  • Ultra low levels of ions and extractables makes it ideal for critical cleaning tasks
Using Contec’s patented Particle Attraction Technology (PAT) system, the wipe attracts andMore Product Information