Wipes, Amplitude™ Sigma™ Plus
Toimittaja: CONTEC
C1E-99 C1E-1212
CONTC1E-99EA 784 EUR
CONTC1E-99 CONTC1E-1212
Wipes, Amplitude™ Sigma™ Plus
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
The Amplitude™ Sigma™ Plus wipes are strong and durable, compatible with most solvents, and have outstanding sorbency.
  • Hydroentangled cellulose/poylester provides excellent strength and durability
  • Low levels of particles and extractables
  • Variety of blended fibers enable excellent absorbing capacity
Amplitude™ Sigma™ Plus Wipes are constructed using a nonwoven fabric that is a hydroentangled blend of polyester and cellulose fibers. Wet or dry, the wipe maintains a high level of strength and durability.
Specially treated to reduce particles andMore Product Information