Cleanroom wipes, Anticon 100® HeavyWeight™
Toimittaja: CONTEC
492226-939 492222-813 492222-815
CONT492226-939EA 1200 EUR
CONT492226-939 CONT492222-813 CONT492222-815
Cleanroom wipes, Anticon 100® HeavyWeight™
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
These wipes are made from a 100% no-run interlock knit polyester construction with knife cut edges.
  • Manufactured from 100% continuous filament polyester in a no-run interlock knit construction
  • Patented textile technology creates a polyester wipe with high sorbency
  • Laundered and packed in a Class 4 cleanroom
  • Lowest fibres and particles available for a cut edge wipe reduces risk of contamination
  • High sorptive capacity decreases the number of wipes required
  • Excellent choice for wiping and absorbing spills in critical cleanroom environments
Anticon 100®More Product Information