Lämpötilaloggeri
Toimittaja: COMARK
N2011 N2012 N2013 N2014
COMAN2011EA 595 EUR
COMAN2011 COMAN2012 COMAN2013 COMAN2014
Lämpötilaloggeri
Dataloggerit
These data loggers measure both humidity and temperature when monitoring food in storage, transit or processing as well as pharmaceutical and general industrial applications.
  • Large memory capacity and single button control
  • Ability to log over multiple periods
  • Windows™ based evolution software for fast data download and analysis
  • LED indication of active logging and temperature or humidity alarm conditions
  • LCD displays on selected models for instant checks on current readings and alarms
  • Additional beeper warning of temperature and humidity alarm conditions