Pump, piston siphon
Toimittaja: NRG DEVELOPMENTS
5900
COLL5900EA 0 EUR
COLL5900
Pump, piston siphon
Pumput Rumpupumput
Piston operated siphon pump. Adjustable bung allows the pump to be used with different containers.
  • With PE suction tube
  • For drums up to 205 litres